accessibility menu, dialog, popup

UserName

CAS RN 1328-51-4

NNNSOOONNSOOONNNNaNaCu

CAS RN 1328-51-4

IUPAC Name: copper(2+) disodium 2,11,20,29,37,38,39,40-octaazanonacyclo[28.6.1.1³,¹⁰.1¹²,¹⁹.1²¹,²⁸.0⁴,⁹.0¹³,¹⁸.0²²,²⁷.0³¹,³⁶]tetraconta-1(36),2,4,6,8,10(40),11,13,15,17,19(39),20,22,24,26,28,30,32,34-nonadecaene-37,38-diide-5,17-disulfonate
Synonyms: copper(2+) disodium 2,11,20,29,37,38,39,40-octaazanonacyclo[28.6.1.1³,¹⁰.1¹²,¹⁹.1²¹,²⁸.0⁴,⁹.0¹³,¹⁸.0²²,²⁷.0³¹,³⁶]tetraconta-1(36),2,4,6,8,10(40),11,13,15,17,19(39),20,22,24,26,28,30,32,34-nonadecaene-37,38-diide-5,17-disulfonate
Molecular Weight (g/mol): 780.16
Molecular Formula: C32H14CuN8Na2O6S2
InChi Key: DKBXPLYSDKSFEQ-UHFFFAOYSA-L
SMILES: [Na+].[Na+].[Cu++].[O-]S(=O)(=O)C1=C2C3=NC(=NC4=NC(=NC5=C6C=CC=CC6=C([N-]5)N=C5[N-]C(=N3)C3=CC=CC=C53)C3=C(C=CC=C43)S([O-])(=O)=O)C2=CC=C1
Quantity 
  • (2)
  • (2)
Color 
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)