accessibility menu, dialog, popup

UserName

CAS RN 13195-64-7

H3CCH3OOOOCH3CH3

CAS RN 13195-64-7

IUPAC Name: 1,3-bis(propan-2-yl) propanedioate
Synonyms: 1,3-diisopropyl propanedioate
Molecular Weight (g/mol): 188.22
Molecular Formula: C9H16O4
InChi Key: QRVSDVDFJFKYKA-UHFFFAOYSA-N
SMILES: CC(C)OC(=O)CC(=O)OC(C)C
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results