accessibility menu, dialog, popup

UserName

CAS RN 131724-45-3

H3CH3CCH3OONH(S)NOO

CAS RN 131724-45-3

IUPAC Name: (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-(4-cyanophenyl)propanoate
Synonyms: (2S)-2-[(tert-butoxycarbonyl)amino]-3-(4-cyanophenyl)propanoate
Molecular Weight (g/mol): 289.31
Molecular Formula: C15H17N2O4
InChi Key: RMBLTLXJGNILPG-LBPRGKRZSA-M
SMILES: CC(C)(C)OC(=O)N[C@@H](CC1=CC=C(C=C1)C#N)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)