accessibility menu, dialog, popup

UserName

CAS RN 1314-18-7

SrOO

CAS RN 1314-18-7

IUPAC Name: strontium(2+) dioxidanediide
Synonyms: strontium peroxide
Molecular Weight (g/mol): 119.62
Molecular Formula: O2Sr
InChi Key: UHCGLDSRFKGERO-UHFFFAOYSA-N
SMILES: [Sr++].[O-][O-]
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results