accessibility menu, dialog, popup

UserName

CAS RN 13083-04-0

NH3NH3NOOHONOOHONOOONOOONOOOH2OH2OH2OH2OCe

CAS RN 13083-04-0

IUPAC Name: cerium(3+) bis(nitric acid) diamine tetrahydrate trinitrate
Synonyms: cerium(3+) bis(nitric acid) diamine tetrahydrate trinitrate
Molecular Weight (g/mol): 558.27
Molecular Formula: CeH16N7O19
InChi Key: AIVZRMAXALRCHB-UHFFFAOYSA-N
SMILES: N.N.O.O.O.O.[Ce+3].O[N+]([O-])=O.O[N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Grade 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results