accessibility menu, dialog, popup

UserName

CAS RN 130663-39-7

H3CNH3CNNNOHOOH3C

CAS RN 130663-39-7

IUPAC Name: 1-{[4-(dimethylamino)-3-methylphenyl]methyl}-5-(2,2-diphenylacetyl)-1H,4H,5H,6H,7H-imidazo[4,5-c]pyridine-6-carboxylic acid
Synonyms: 1-{[4-(dimethylamino)-3-methylphenyl]methyl}-5-(2,2-diphenylacetyl)-4H,6H,7H-imidazo[4,5-c]pyridine-6-carboxylic acid
Molecular Weight (g/mol): 508.62
Molecular Formula: C31H32N4O3
InChi Key: YSTVFDAKLDMYCR-UHFFFAOYNA-N
SMILES: CN(C)C1=CC=C(CN2C=NC3=C2CC(N(C3)C(=O)C(C2=CC=CC=C2)C2=CC=CC=C2)C(O)=O)C=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results