CAS RN 129-06-6
CAS RN 129-06-6
IUPAC Name:
sodium 2,4-dioxo-3-(3-oxo-1-phenylbutyl)-3,4-dihydro-2H-1-benzopyran-3-ide
Synonyms:
warfarin sodium
Molecular Weight (g/mol):
330.32
Molecular Formula:
C19H15NaO4
InChi Key:
PAIQCHHPKBVDOM-UHFFFAOYNA-N
SMILES:
[Na+].CC(=O)CC([C-]1C(=O)OC2=CC=CC=C2C1=O)C1=CC=CC=C1
1
–
1
of
1
results
1
(±)-Warfarin Sodium, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.