CAS RN 128495-46-5
CAS RN 128495-46-5
IUPAC Name:
4-fluoro-3-methoxybenzaldehyde
Synonyms:
4-fluoro-3-methoxybenzaldehyde
Molecular Weight (g/mol):
154.14
Molecular Formula:
C8H7FO2
InChi Key:
NALVGTOMKSKFFV-UHFFFAOYSA-N
SMILES:
COC1=CC(C=O)=CC=C1F
1
–
1
of
1
results
1
4-Fluoro-3-methoxybenzaldehyde, 98%
CAS: 128495-46-5 Molecular Formula: C8H7FO2 Molecular Weight (g/mol): 154.14 MDL Number: MFCD00143320 InChI Key: NALVGTOMKSKFFV-UHFFFAOYSA-N Synonym: 3-methoxy-4-fluorobenzaldehyde,4-fluoro-m-anisaldehyde,4-fluoro-3-methoxy-benzaldehyde,benzaldehyde, 4-fluoro-3-methoxy,pubchem1449,acmc-209bf6,ksc490m2d,3-methoxy-4-fluoro benzaldehyde,3-methoxy-4-fluoro-benzaldehyde,nalvgtomkskffv-uhfffaoysa PubChem CID: 2737358 IUPAC Name: 4-fluoro-3-methoxybenzaldehyde SMILES: COC1=CC(C=O)=CC=C1F