CAS RN 1268340-94-8
IUPAC Name:
trifluoro[(morpholin-4-ium-4-yl)methyl]boranuide
Synonyms:
trifluoro(morpholin-4-ium-4-ylmethyl)boranuide
Molecular Weight (g/mol):
168.95
Molecular Formula:
C5H11BF3NO
InChi Key:
FQKLIGKQQGNVHW-UHFFFAOYSA-O
SMILES:
F[B-](F)(F)C[NH+]1CCOCC1
1
–
1
of
1
results
1
(Morpholinium-4-ylmethyl)trifluoroborate internal salt, 95%
CAS: 1268340-94-8 Molecular Formula: C5H11BF3NO Molecular Weight (g/mol): 168.954 MDL Number: MFCD20134168 InChI Key: FQKLIGKQQGNVHW-UHFFFAOYSA-O Synonym: trifluoro morpholin-4-ium-4-ylmethyl boranuide,morpholinium-4-ylmethyl trifluoroborate internal salt,morpholinium-4-yl-methyl trifluoroborate internal salt,morpholinium-4-ylmethyltrifluoroboronanion,morpholin-4-ium-4-yl methyl trifluoroborate,trifluoro morpholin-4-ium-4-yl methyl boranuide,trifluoro morpholin-4-ium-4-yl methyl borate 1- PubChem CID: 53243644 IUPAC Name: trifluoro(morpholin-4-ium-4-ylmethyl)boranuide SMILES: [B-](C[NH+]1CCOCC1)(F)(F)F