accessibility menu, dialog, popup

UserName

CAS RN 124700-41-0

IOHOF

CAS RN 124700-41-0

IUPAC Name: 2-fluoro-5-iodobenzoic acid
Synonyms: 2-fluoro-5-iodobenzoic acid
Molecular Weight (g/mol): 266.01
Molecular Formula: C7H4FIO2
InChi Key: QNNJHBNTHVHALE-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(F)C=CC(I)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results