CAS RN 1246819-32-8
CAS RN 1246819-32-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Dehydro Mefloquine-d5, TRC
CAS: 1246819-32-8 Molecular Formula: C17H5D5F6N2O Molecular Weight (g/mol): 377.3 Synonym: alpha-2-Pyridinyl-2,8-bis(trifluoromethyl)-4-quinolinemethanol-d5; SMILES: [2H]c1nc(c([2H])c([2H])c1[2H])C([2H])(O)c2cc(nc3c(cccc23)C(F)(F)F)C(F)(F)F