CAS RN 1246818-75-6
CAS RN 1246818-75-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
rac-1-[2-(Cyano)phenyl]-3-O-methoxymethyl-3-piperidinol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
rac-1-[2-(Cyano)phenyl]-3-O-methoxymethyl-3-piperidinol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.