CAS RN 124-26-5
CAS RN 124-26-5
IUPAC Name:
octadecanamide
Synonyms:
stearamide
Molecular Weight (g/mol):
283.5
Molecular Formula:
C18H37NO
InChi Key:
LYRFLYHAGKPMFH-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCCCCCCCC(N)=O
1
–
2
of
2
results
1
Octadecanamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Octadecanamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.