CAS RN 1239233-87-4
CAS RN 1239233-87-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-[3-Carboxy-4-(2-methylpropoxy)phenyl]-4-methyl-5-thiazolecarboxylic Acid, TRC
CAS: 1239233-87-4 Molecular Formula: C16H17NO5S Molecular Weight (g/mol): 335.37 Synonym: Febuxostat Impurity IUPAC Name: 2-[3-carboxy-4-(2-methylpropoxy)phenyl]-4-methyl-1,3-thiazole-5-carboxylic acid SMILES: OC(C(S1)=C(C)N=C1C2=CC(C(O)=O)=C(OCC(C)C)C=C2)=O