CAS RN 123447-62-1
CAS RN 123447-62-1
IUPAC Name:
6-fluoro-1-methyl-7-{4-[(5-methyl-2-oxo-2H-1,3-dioxol-4-yl)methyl]piperazin-1-yl}-4-oxo-1H,4H-[1,3]thiazeto[3,2-a]quinoline-3-carboxylic acid
Synonyms:
prulifloxacin
Molecular Weight (g/mol):
461.46
Molecular Formula:
C21H20FN3O6S
InChi Key:
PWNMXPDKBYZCOO-UHFFFAOYNA-N
SMILES:
CC1SC2=C(C(O)=O)C(=O)C3=CC(F)=C(C=C3N12)N1CCN(CC2=C(C)OC(=O)O2)CC1
1
–
3
of
3
results
1
Prulifloxacin, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Prulifloxacin, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Prulifloxacin, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.