accessibility menu, dialog, popup

UserName

N(R)N(R)O(R)(R)NNFNH2HOHOCl

CAS RN 123318-82-1

IUPAC Name: (2R,3R,4S,5R)-5-(6-amino-2-chloro-9H-purin-9-yl)-4-fluoro-2-(hydroxymethyl)oxolan-3-ol
Synonyms: clofarabine evoltra cl-F-ara-A clolar C1-F-ara-A clofarabine (product) clofarex genzyme brand of clofarabine clofarabine (substance) CAFdA
Molecular Weight (g/mol): 303.68
Molecular Formula: C10H11ClFN5O3
InChi Key: WDDPHFBMKLOVOX-AYQXTPAHSA-N
SMILES: NC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@@H]3F)C2=NC(Cl)=N1
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (2)
  • (3)
  • (3)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)