accessibility menu, dialog, popup

UserName

H2OClClH3CONOHN(R)(R)SNOHOONH3CSNH2NH(Z)

CAS RN 123171-59-5

IUPAC Name: hydrogen 1-{[(6R,7R)-7-[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-(methoxyimino)acetamido]-2-carboxy-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-en-3-yl]methyl}-1-methylpyrrolidin-1-ium hydrate dichloride
Synonyms: hydrogen cefepime(1+) hydrate dichloride
Molecular Weight (g/mol): 571.49
Molecular Formula: C19H28Cl2N6O6S2
InChi Key: LRAJHPGSGBRUJN-OMIVUECESA-N
SMILES: [H+].O.[Cl-].[Cl-].CO\N=C(/C(=O)N[C@H]1[C@H]2SCC(C[N+]3(C)CCCC3)=C(N2C1=O)C(O)=O)C1=CSC(N)=N1
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)
Color 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)