accessibility menu, dialog, popup

UserName

CAS RN 123-46-6

ClCH3NCH3H3CONHH2N

CAS RN 123-46-6

IUPAC Name: [(hydrazinecarbonyl)methyl]trimethylazanium chloride
Synonyms: [(hydrazinecarbonyl)methyl]trimethylazanium chloride
Molecular Weight (g/mol): 167.64
Molecular Formula: C5H14ClN3O
InChi Key: YSULOORXQBDPCU-UHFFFAOYSA-N
SMILES: [Cl-].C[N+](C)(C)CC(=O)NN

 No Refinements