CAS RN 1228445-38-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
STEMCELL Technologies Pyrintegrin
Integrin and tyrosine kinase activator; Activates Integrin, FGFR, IGFR, and HER2