accessibility menu, dialog, popup

UserName

CAS RN 122308-25-2

H3COOSOOCH3

CAS RN 122308-25-2

IUPAC Name: 1,3-dimethyl 2-[(thiophen-2-yl)methyl]propanedioate
Synonyms: 1,3-dimethyl 2-(thiophen-2-ylmethyl)propanedioate
Molecular Weight (g/mol): 228.26
Molecular Formula: C10H12O4S
InChi Key: PIIHAJHYNTWOAF-UHFFFAOYSA-N
SMILES: COC(=O)C(CC1=CC=CS1)C(=O)OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results