accessibility menu, dialog, popup

UserName

H3CNHOCH3OCH3

CAS RN 122-07-6

IUPAC Name: (2,2-dimethoxyethyl)(methyl)amine
Synonyms: (2,2-dimethoxyethyl)(methyl)amine
Molecular Weight (g/mol): 119.16
Molecular Formula: C5H13NO2
InChi Key: HUMIEJNVCICTPJ-UHFFFAOYSA-N
SMILES: CNCC(OC)OC
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results