accessibility menu, dialog, popup

UserName

CAS RN 12163-26-7

ONbOOONbOOMg

CAS RN 12163-26-7

IUPAC Name: diniobium(5+) dimagnesium(2+) heptaoxidandiide
Synonyms: diniobium(5+) dimagnesium(2+) heptaoxidandiide
Molecular Weight (g/mol): 346.42
Molecular Formula: Mg2Nb2O7
InChi Key: CWMQWEHVVUJOBW-UHFFFAOYSA-N
SMILES: [O--].[O--].[O--].[O--].[O--].[O--].[O--].[Mg++].[Mg++].[Nb+5].[Nb+5]
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results