accessibility menu, dialog, popup

UserName

CAS RN 12142-88-0

NiTe

CAS RN 12142-88-0

IUPAC Name: tellanylidenenickel
Synonyms: nickel telluride
Molecular Weight (g/mol): 186.29
Molecular Formula: NiTe
InChi Key: NPEUSMKUOOTUGX-UHFFFAOYSA-N
SMILES: [Ni]=[Te]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results