CAS RN 12141-67-2
IUPAC Name:
hexasodium tris(dioxotungstenbis(olate)) nonakis(trioxotungsten)
Synonyms:
hexasodium nonakis(tungsten trioxide) tritungstate
Molecular Weight (g/mol):
2967.98
Molecular Formula:
Na6O39W12
InChi Key:
AZCSOJKJFMWYCX-UHFFFAOYSA-N
SMILES:
[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.[O-][W]([O-])(=O)=O.[O-][W]([O-])(=O)=O.[O-][W]([O-])(=O)=O
1
–
3
of
3
results
1
Sodium metatungstate, 2.82 density solution in water, stabilized
CAS: 12141-67-2 Molecular Formula: H2Na6O40W12 MDL Number: MFCD00150663 Synonym: Sodium polytungstate
2
Sodium Metatungstate Hydrate, TRC
CAS: 12141-67-2 Molecular Formula: O39Na6W12•H2O Molecular Weight (g/mol): 2986.01 Synonym: Sodium metatungstate (Na6H2W12O40),Sodium tungstate (Na6H2W12O40),Sodium tungstate(VI) (Na6H2W12O40),Tungstate (W12(OH)2O386-) Hexasodium,Tungstic Acid (H8W12O40) Hexasodium Salt,SPT,POM 1 IUPAC Name: hexasodium;dihydroxy(dioxo)tungsten;dioxido(dioxo)tungsten;trioxotungsten SMILES: OC(=O)c1ccc(cc1)NC[C@@H]2N(C=O)C3=C(NC(N)=NC3=O)NC2