accessibility menu, dialog, popup

UserName

OHPOOHOHPOOHONH2H2OH2OH2ONa

CAS RN 121268-17-5

IUPAC Name: sodium trihydrate hydrogen (4-amino-1-hydroxy-1-phosphonobutyl)phosphonate
Synonyms: arendal alendros adronat
Molecular Weight (g/mol): 325.12
Molecular Formula: C4H18NNaO10P2
InChi Key: DCSBSVSZJRSITC-UHFFFAOYNA-M
SMILES: O.O.O.[Na+].NCCCC(O)(P(O)(O)=O)P(O)([O-])=O
Molecular Weight (g/mol) 
  • (4)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results