accessibility menu, dialog, popup

UserName

CAS RN 121-90-4

ONOClO

CAS RN 121-90-4

IUPAC Name: 3-nitrobenzoyl chloride
Synonyms: benzoyl chloride, 3-nitro-
Molecular Weight (g/mol): 185.56
Molecular Formula: C7H4ClNO3
InChi Key: NXTNASSYJUXJDV-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC(=C1)C(Cl)=O
Boiling Point 
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)