accessibility menu, dialog, popup

UserName

CAS RN 121-01-7

NH2FFFNOO

CAS RN 121-01-7

IUPAC Name: 4-nitro-2-(trifluoromethyl)aniline
Synonyms: 4-nitro-2-(trifluoromethyl)aniline
Molecular Weight (g/mol): 206.12
Molecular Formula: C7H5F3N2O2
InChi Key: HOTZLWVITTVZGY-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1C(F)(F)F)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)