accessibility menu, dialog, popup

UserName

CAS RN 12069-00-0

SePb

CAS RN 12069-00-0

IUPAC Name: plumbaneselone
Synonyms: lead selenide
Molecular Weight (g/mol): 286.2
Molecular Formula: PbSe
InChi Key: GGYFMLJDMAMTAB-UHFFFAOYSA-N
SMILES: [Se]=[Pb]
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results