accessibility menu, dialog, popup

UserName

CAS RN 12060-01-4

OZrOOPb

CAS RN 12060-01-4

IUPAC Name: λ²-lead(2+) oxozirconiumbis(olate)
Synonyms: λ²-lead(2+) oxozirconiumbis(olate)
Molecular Weight (g/mol): 346.4
Molecular Formula: O3PbZr
InChi Key: RSCSNKLWUCUPDE-UHFFFAOYSA-N
SMILES: [Pb++].[O-][Zr]([O-])=O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)