CAS RN 12053-12-2
CAS RN 12053-12-2
IUPAC Name:
chromium(3+) antimony
Synonyms:
chromium(3+) antimony
Molecular Weight (g/mol):
173.75
Molecular Formula:
CrSb
InChi Key:
BUNMATGODDVOLM-UHFFFAOYSA-N
SMILES:
[Cr+3].[Sb]
1
–
1
of
1
results
1
Chromium(III) antimonide, 99%, Thermo Scientific™
CAS: 12053-12-2 Molecular Formula: CrSb Molecular Weight (g/mol): 173.756 MDL Number: MFCD00049884 InChI Key: KECAIQNCFZTEBM-UHFFFAOYSA-N Synonym: chromiumylidynestibane,chromium iii antimonide,chromium antimonide, PubChem CID: 24884257 IUPAC Name: stibanylidynechromium SMILES: [Cr]#[Sb]