CAS RN 12036-22-5
CAS RN 12036-22-5
IUPAC Name:
dioxotungsten
Synonyms:
tungsten(IV) oxide
Molecular Weight (g/mol):
215.84
Molecular Formula:
O2W
InChi Key:
DZKDPOPGYFUOGI-UHFFFAOYSA-N
SMILES:
O=[W]=O
1
–
1
of
1
results
1
Tungsten(IV) oxide, 99.9% (metals basis)
CAS: 12036-22-5 Molecular Formula: O2W Molecular Weight (g/mol): 215.84 MDL Number: MFCD00049709 InChI Key: DZKDPOPGYFUOGI-UHFFFAOYSA-N Synonym: tungsten dioxide,tungsten iv oxide,tungsten oxide wo2,acmc-1bqvs,tungsten iv oxide-w,tungsten iv oxide trace metals basis 5g,tungsten iv oxide,-100 mesh trace metals basis PubChem CID: 82850 IUPAC Name: dioxotungsten SMILES: O=[W]=O