accessibility menu, dialog, popup

UserName

CAS RN 120351-90-8

H3NOF

CAS RN 120351-90-8

IUPAC Name: 2-(2-fluorophenoxy)ethan-1-aminium
Synonyms: 2-(2-fluorophenoxy)ethanaminium
Molecular Weight (g/mol): 156.18
Molecular Formula: C8H11FNO
InChi Key: LXOQYJMQPRGSTL-UHFFFAOYSA-O
SMILES: [NH3+]CCOC1=CC=CC=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results