accessibility menu, dialog, popup

UserName

CAS RN 12007-98-6

BBBBBWW

CAS RN 12007-98-6

IUPAC Name: pentaboron ditungsten
Synonyms: pentaboron ditungsten
Molecular Weight (g/mol): 421.73
Molecular Formula: B5W2
InChi Key: CXOKLXRCTOUPEV-UHFFFAOYSA-N
SMILES: [B].[B].[B].[B].[B].[W].[W]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results