accessibility menu, dialog, popup

UserName

CAS RN 119-65-3

N

CAS RN 119-65-3

IUPAC Name: isoquinoline
Synonyms: isoquinoline benzo(c)pyridine leucoline β-quinoline isochinolin 3,4-benzopyridine
Molecular Weight (g/mol): 129.16
Molecular Formula: C9H7N
InChi Key: AWJUIBRHMBBTKR-UHFFFAOYSA-N
SMILES: C1=CC=C2C=NC=CC2=C1
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)