accessibility menu, dialog, popup

UserName

OFFFH2NF

CAS RN 116369-23-4

IUPAC Name: 2-fluoro-5-(trifluoromethoxy)aniline
Synonyms: 2-fluoro-5-(trifluoromethoxy)aniline
Molecular Weight (g/mol): 195.12
Molecular Formula: C7H5F4NO
InChi Key: HQUWXEXDIIWBBY-UHFFFAOYSA-N
SMILES: NC1=C(F)C=CC(OC(F)(F)F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results