accessibility menu, dialog, popup

UserName

CAS RN 115-09-3

H3CHgCl

CAS RN 115-09-3

IUPAC Name: chloro(methyl)mercury
Synonyms: methylmercury chloride chloromethylmercury mercury methyl chloride methylmercuric chloride MeHgCl
Molecular Weight (g/mol): 251.08
Molecular Formula: CH3ClHg
InChi Key: BABMCXWQNSQAOC-UHFFFAOYSA-M
SMILES: C[Hg]Cl
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results