CAS RN 114873-13-1
CAS RN 114873-13-1
IUPAC Name:
(2R)-2-{[(tert-butoxy)carbonyl]amino}-3-(3,4-dichlorophenyl)propanoate
Synonyms:
(2R)-2-[(tert-butoxycarbonyl)amino]-3-(3,4-dichlorophenyl)propanoate
Molecular Weight (g/mol):
333.19
Molecular Formula:
C14H16Cl2NO4
InChi Key:
UGZIQCCPEDCGGN-LLVKDONJSA-M
SMILES:
CC(C)(C)OC(=O)N[C@H](CC1=CC=C(Cl)C(Cl)=C1)C([O-])=O
1
–
1
of
1
results
1
N-Boc-3,4-dichloro-D-phenylalanine, 95%, Thermo Scientific™
CAS: 114873-13-1 Molecular Formula: C14H16Cl2NO4 Molecular Weight (g/mol): 333.19 MDL Number: MFCD00151887 InChI Key: UGZIQCCPEDCGGN-LLVKDONJSA-M Synonym: boc-3,4-dichloro-d-phenylalanine,boc-d-phe 3,4-cl2-oh,r-2-tert-butoxycarbonyl amino-3-3,4-dichlorophenyl propanoic acid,boc-d-3,4-dichlorophe,boc-d-3,4-dichlorophenylalanine,2r-2-tert-butoxycarbonyl amino-3-3,4-dichlorophenyl propanoic acid,boc-d-phe 3,4-dicl-oh,boc-3,4-dichloro-d-phe-oh,n-boc-d-3,4-dichlorophenylalanine,d-phenylalanine, 3,4-dichloro-n-1,1-dimethylethoxy carbonyl PubChem CID: 7015811 SMILES: CC(C)(C)OC(=O)N[C@H](CC1=CC=C(Cl)C(Cl)=C1)C([O-])=O