accessibility menu, dialog, popup

UserName

CAS RN 1147-56-4

ONHNSN(Z)

CAS RN 1147-56-4

IUPAC Name: (1Z)-1-[2-(1,3-thiazol-2-yl)hydrazin-1-ylidene]-1,2-dihydronaphthalen-2-one
Synonyms: (1Z)-1-[2-(1,3-thiazol-2-yl)hydrazin-1-ylidene]naphthalen-2-one
Molecular Weight (g/mol): 255.3
Molecular Formula: C13H9N3OS
InChi Key: GCBAKGGJQPMJLL-QINSGFPZSA-N
SMILES: O=C1C=CC2=CC=CC=C2\C1=N\NC1=NC=CS1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results