CAS RN 114456-61-0
IUPAC Name:
trisodium 2-hydroxypropane-1,2,3-tricarboxylate hydrate
Synonyms:
trisodium citrate hydrate
Molecular Weight (g/mol):
276.08
Molecular Formula:
C6H7Na3O8
InChi Key:
PKIDNTKRVKSLDB-UHFFFAOYSA-K
SMILES:
O.[Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
1
–
1
of
1
results
1
Citric acid, trisodium salt hydrate, 99.8%, for molecular biology, DNAse, RNAse and Protease free
CAS: 114456-61-0 Molecular Formula: C6H5Na3O7 Molecular Weight (g/mol): 258.07 MDL Number: MFCD00150032 InChI Key: HRXKRNGNAMMEHJ-UHFFFAOYSA-K Synonym: trisodium citrate hydrate,sodium citrate tribasic hydrate,citric acid trisodium salt hydrate,sodium citrate, tribasic,sodium citrate monohydrate,c6h5o7.3na.h2o,sodium citrate tribasic hydrate, reagentplus r,trisodium 2-hydroxypropane-1,2,3-tricarboxylate hydrate PubChem CID: 21868242 SMILES: [Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O