CAS RN 1141931-86-3
CAS RN 1141931-86-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
1,4-Bis[(1S)-1-(4-methoxyphenyl)ethyl]-2,5-piperazinedione, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1,4-Bis[(1S)-1-(4-methoxyphenyl)ethyl]-2,5-piperazinedione, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
1,4-Bis[(1S)-1-(4-methoxyphenyl)ethyl]-2,5-piperazinedione, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.