CAS RN 113798-74-6
IUPAC Name:
2,3,6-trifluorophenol
Synonyms:
2,3,6-trifluorophenol
Molecular Weight (g/mol):
148.08
Molecular Formula:
C6H3F3O
InChi Key:
QSFGUSFDWCVXNR-UHFFFAOYSA-N
SMILES:
OC1=C(F)C=CC(F)=C1F
1
–
1
of
1
results
1
2,3,6-Trifluorophenol, 98%
CAS: 113798-74-6 Molecular Formula: C6H3F3O Molecular Weight (g/mol): 148.08 MDL Number: MFCD00061214 InChI Key: QSFGUSFDWCVXNR-UHFFFAOYSA-N PubChem CID: 517988 IUPAC Name: 2,3,6-trifluorophenol SMILES: OC1=C(F)C=CC(F)=C1F