accessibility menu, dialog, popup

UserName

CAS RN 1134156-31-2

OONNNNH2NHNClClHOHON

CAS RN 1134156-31-2

IUPAC Name: 4-({[5-(6-amino-8-{[(3,4-dichlorophenyl)methyl]amino}-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}methyl)benzonitrile
Synonyms: 4-({[5-(6-amino-8-{[(3,4-dichlorophenyl)methyl]amino}purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}methyl)benzonitrile
Molecular Weight (g/mol): 556.4
Molecular Formula: C25H23Cl2N7O4
InChi Key: ZXGGCBQORXDVTE-UHFFFAOYNA-N
SMILES: NC1=C2N=C(NCC3=CC(Cl)=C(Cl)C=C3)N(C3OC(COCC4=CC=C(C=C4)C#N)C(O)C3O)C2=NC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results