accessibility menu, dialog, popup

UserName

CAS RN 113209-68-0

OOF

CAS RN 113209-68-0

IUPAC Name: 7-fluoro-3,4-dihydro-2H-1-benzopyran-4-one
Synonyms: 7-fluoro-2,3-dihydro-1-benzopyran-4-one
Molecular Weight (g/mol): 166.15
Molecular Formula: C9H7FO2
InChi Key: HRPULQFHSZKTNA-UHFFFAOYSA-N
SMILES: FC1=CC2=C(C=C1)C(=O)CCO2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results