CAS RN 1127442-82-3
CAS RN 1127442-82-3
IUPAC Name:
4-{3,5-dioxo-4-azatricyclo[5.2.1.0²,⁶]dec-8-en-4-yl}-N-(quinolin-8-yl)benzamide
Synonyms:
4-{3,5-dioxo-4-azatricyclo[5.2.1.0²,⁶]dec-8-en-4-yl}-N-(quinolin-8-yl)benzamide
Molecular Weight (g/mol):
409.45
Molecular Formula:
C25H19N3O3
InChi Key:
ZGSXEXBYLJIOGF-UHFFFAOYNA-N
SMILES:
O=C(NC1=C2N=CC=CC2=CC=C1)C1=CC=C(C=C1)N1C(=O)C2C3CC(C=C3)C2C1=O
1
–
3
of
3
results
1
Tocris Bioscience™ endo-IWR 1
CAS: 1127442-82-3 Molecular Formula: C25H19N3O3
2
Tocris Bioscience™ endo-IWR 1
endo-IWR 1 synthesized to Ancillary Material Grade
3