accessibility menu, dialog, popup

UserName

NHSNH2

CAS RN 112401-09-9

IUPAC Name: piperidine-4-carbothioamide
Synonyms: piperidine-4-carbothioamide
Molecular Weight (g/mol): 144.24
Molecular Formula: C6H12N2S
InChi Key: LXOBYAWHAIZPPU-UHFFFAOYSA-N
SMILES: NC(=S)C1CCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results