CAS RN 109919-26-8
IUPAC Name:
2-bromo-6-chloro-4-(trifluoromethyl)aniline
Synonyms:
2-bromo-6-chloro-4-(trifluoromethyl)aniline
Molecular Weight (g/mol):
274.47
Molecular Formula:
C7H4BrClF3N
InChi Key:
OVJCOPVKCCJSFE-UHFFFAOYSA-N
SMILES:
NC1=C(Cl)C=C(C=C1Br)C(F)(F)F
1
–
1
of
1
results
1
2-Bromo-6-chloro-4-(trifluoromethyl)aniline, 98%, Thermo Scientific™
CAS: 109919-26-8 Molecular Formula: C7H4BrClF3N Molecular Weight (g/mol): 274.47 MDL Number: MFCD00042180 InChI Key: OVJCOPVKCCJSFE-UHFFFAOYSA-N Synonym: 2-bromo-6-chloro-4-trifluoromethyl aniline,4-amino-3-bromo-5-chlorobenzotrifluoride,buttpark 44\03-53,2-bromo-6-chloro-4-trifluoromethyl phenylamine,pubchem2921,ksc498c4n,2-bromo-6-chloro-4-trifluoromethyl benzenamine,benzenamine, 2-bromo-6-chloro-4-trifluoromethyl PubChem CID: 737196 IUPAC Name: 2-bromo-6-chloro-4-(trifluoromethyl)aniline SMILES: NC1=C(Cl)C=C(C=C1Br)C(F)(F)F