accessibility menu, dialog, popup

UserName

CAS RN 1092460-81-5

FOOFFF

CAS RN 1092460-81-5

IUPAC Name: 5-fluoro-2-(trifluoromethoxy)benzaldehyde
Synonyms: 5-fluoro-2-(trifluoromethoxy)benzaldehyde
Molecular Weight (g/mol): 208.11
Molecular Formula: C8H4F4O2
InChi Key: DLJQIFHQYJCQHB-UHFFFAOYSA-N
SMILES: FC1=CC(C=O)=C(OC(F)(F)F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results