accessibility menu, dialog, popup

UserName

CAS RN 1092351-67-1

H3CCH3NNOHNHNNNH2(E/Z)

CAS RN 1092351-67-1

IUPAC Name: 2-[4-amino-1-(propan-2-yl)-1H,2H,3H-pyrazolo[3,4-d]pyrimidin-3-ylidene]-2H-indol-5-ol
Synonyms: 2-{4-amino-1-isopropyl-2H-pyrazolo[3,4-d]pyrimidin-3-ylidene}indol-5-ol
Molecular Weight (g/mol): 308.35
Molecular Formula: C16H16N6O
InChi Key: MTICIDWIKCANDD-UHFFFAOYSA-N
SMILES: CC(C)N1NC(=C2C=C3C=C(O)C=CC3=N2)C2=C1N=CN=C2N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results