CAS RN 107756-30-9
CAS RN 107756-30-9
IUPAC Name:
N-[5-(4,5-dihydro-1H-imidazol-2-yl)-2-hydroxy-5,6,7,8-tetrahydronaphthalen-1-yl]methanesulfonamide hydrobromide
Synonyms:
N-[5-(4,5-dihydro-1H-imidazol-2-yl)-2-hydroxy-5,6,7,8-tetrahydronaphthalen-1-yl]methanesulfonamide hydrobromide
Molecular Weight (g/mol):
390.3
Molecular Formula:
C14H20BrN3O3S
InChi Key:
LRFLWCZMTGTUEP-UHFFFAOYNA-N
SMILES:
Br.CS(=O)(=O)NC1=C(O)C=CC2=C1CCCC2C1=NCCN1
1
–
1
of
1
results
1
A 61603 hydrobromide, Tocris Bioscience™
CAS: 107756-30-9 Molecular Formula: C14H20BrN3O3S Molecular Weight (g/mol): 390.296 InChI Key: LRFLWCZMTGTUEP-UHFFFAOYSA-N Synonym: a 61603 hydrobromide,n-5-4,5-dihydro-1h-imidazol-2-yl-2-hydroxy-5,6,7,8-tetrahydronaphthalen-1-yl methanesulfonamide hydrobromide PubChem CID: 9865178 IUPAC Name: N-[5-(4,5-dihydro-1H-imidazol-2-yl)-2-hydroxy-5,6,7,8-tetrahydronaphthalen-1-yl]methanesulfonamide;hydrobromide SMILES: CS(=O)(=O)NC1=C(C=CC2=C1CCCC2C3=NCCN3)O.Br