accessibility menu, dialog, popup

UserName

BOOH3CCH3H3CH3CNNSOO

CAS RN 1073372-04-9

IUPAC Name: 1-(benzenesulfonyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Synonyms: 1-(benzenesulfonyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole
Molecular Weight (g/mol): 334.2
Molecular Formula: C15H19BN2O4S
InChi Key: NPRHNPBHUVCHKP-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CN(N=C1)S(=O)(=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results